Assay Details
| Gene Symbol: C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013484.2 | NP_038512.2 | 5 - 6 | 848 | 62 | |
| GenBank mRNA | AK146812.1 | - | 5 - 6 | 849 | 62 | |
| BC011086.1 | - | 5 - 6 | 811 | 3992881 | 62 | |
| M57891.1 | - | 5 - 6 | 795 | 62 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C2
Genomic Map