Assay Details
| Gene Symbol: Cfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142706.1 | NP_001136178.1 | 12 - 13 | 2022 | 99 | |
| NM_008198.2 | NP_032224.2 | 12 - 13 | 2022 | 99 | ||
| GenBank mRNA | AK098094.1 | - | 12 - 13 | 1682 | 99 | |
| AK149531.1 | - | 12 - 13 | 1655 | 99 | ||
| AK149573.1 | - | 12 - 13 | 1681 | 99 | ||
| AK151220.1 | - | 6 - 7 | 641 | 99 | ||
| AK151618.1 | - | 6 - 7 | 641 | 99 | ||
| AK151974.1 | - | 6 - 7 | 641 | 99 | ||
| BC005451.1 | - | 12 - 13 | 1656 | 3591669 | 99 | |
| K01496.1 | - | 7 - 8 | 762 | 99 | ||
| K01498.1 | - | 2 - 3 | 186 | 99 | ||
| M57890.1 | - | 12 - 13 | 1725 | 99 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cfb
Genomic Map