Assay Details
| Gene Symbol: Cfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142706.1 | NP_001136178.1 | 6 - 7 | 1296 | 113 | |
| NM_008198.2 | NP_032224.2 | 6 - 7 | 1296 | 113 | ||
| GenBank mRNA | AK098094.1 | - | 6 - 7 | 956 | 113 | |
| AK149531.1 | - | 6 - 7 | 929 | 113 | ||
| AK149573.1 | - | 6 - 7 | 955 | 113 | ||
| AK151246.1 | - | 6 - 7 | 1030 | 113 | ||
| BC005451.1 | - | 6 - 7 | 930 | 3591669 | 113 | |
| M57890.1 | - | 6 - 7 | 999 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cfb
Genomic Map