Assay Details
| Gene Symbol: Aif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019467.2 | NP_062340.1 | 3 - 4 | 336 | 59 | |
| GenBank mRNA | AB013745.1 | - | 1 - 2 | 63 | 59 | |
| AF074959.1 | - | 1 - 2 | 74 | 59 | ||
| AK006184.1 | - | 3 - 4 | 336 | 59 | ||
| AK006562.1 | - | 3 - 4 | 337 | 59 | ||
| BC021539.1 | - | 1 - 2 | 204 | 5346103 | 59 | |
| D86382.1 | - | 1 - 2 | 99 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB013745.1, AF074959.1, AK006184.1, AK006562.1, BC021539.1, D86382.1, NM_019467.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Aif1
Genomic Map