Assay Details
| Gene Symbol: Msh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF107352.1 | - | 14 - 15 | 1250 | 73 | |
| AF146227.1 | - | 13 - 14 | 1458 | 73 | ||
| AK132626.1 | - | 1 - 2 | 66 | 73 | ||
| BC141113.1 | - | 13 - 14 | 1378 | 9055725 | 73 | |
| BC145406.1 | - | 13 - 14 | 1378 | 73 | ||
| BC145407.1 | - | 13 - 14 | 1378 | 73 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Msh5
Genomic Map