Assay Details
| Gene Symbol: Msh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF107352.1 | - | 20 - 21 | 1997 | 64 | |
| AF146227.1 | - | 19 - 20 | 2205 | 64 | ||
| AK132626.1 | - | 7 - 8 | 1066 | 64 | ||
| BC141113.1 | - | 19 - 20 | 2125 | 9055725 | 64 | |
| BC145406.1 | - | 19 - 20 | 2115 | 64 | ||
| BC145407.1 | - | 19 - 20 | 2027 | 64 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Msh5
Genomic Map