Assay Details
| Gene Symbol: Msh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF107352.1 | - | 8 - 9 | 717 | 96 | |
| AF146227.1 | - | 7 - 8 | 925 | 96 | ||
| BC141113.1 | - | 7 - 8 | 845 | 9055725 | 96 | |
| BC145406.1 | - | 7 - 8 | 845 | 96 | ||
| BC145407.1 | - | 7 - 8 | 845 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Msh5
Genomic Map