Assay Details
| Gene Symbol: Lsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110101.2 | NP_001103571.1 | 4 - 5 | 392 | 106 | |
| NM_001204273.1 | NP_001191202.1 | 5 - 6 | 427 | 106 | ||
| NM_030597.3 | NP_085100.1 | 4 - 5 | 398 | 106 | ||
| GenBank mRNA | AF204146.1 | - | 4 - 5 | 369 | 106 | |
| AF204147.1 | - | 4 - 5 | 369 | 106 | ||
| AF204148.1 | - | 4 - 5 | 369 | 106 | ||
| AF204149.1 | - | 4 - 5 | 369 | 106 | ||
| AF204150.1 | - | 4 - 5 | 369 | 106 | ||
| AF204151.1 | - | 4 - 5 | 369 | 106 | ||
| AF204152.1 | - | 4 - 5 | 369 | 106 | ||
| AF204153.1 | - | 3 - 4 | 611 | 106 | ||
| AF204154.1 | - | 5 - 6 | 398 | 106 | ||
| AF204155.1 | - | 5 - 6 | 398 | 106 | ||
| AF204157.1 | - | 4 - 5 | 236 | 106 | ||
| AK151605.1 | - | 4 - 5 | 366 | 106 | ||
| AK178855.1 | - | 4 - 5 | 180 | 106 | ||
| BB848823.1 | - | 4 - 5 | 398 | 106 | ||
| BC014288.1 | - | 4 - 5 | 377 | 3982410 | 106 | |
| BC049543.1 | - | 391 | 6774564 | 106 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156879 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156874 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156876 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156873 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156880 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156878 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156877 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2023188 | BB848823.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2023188 | BC049543.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2023188 | AF204153.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2023188 | AF204154.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2023188 | AF204155.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2023181 | BC014288.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2023181 | BC049543.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2023181 | AF204146.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2023181 | AF204147.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2023181 | AF204148.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 519049 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 890206 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161747 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2023181 | BC014288.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2023181 | BC049543.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2023181 | AF204146.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2023181 | AF204147.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2023181 | AF204148.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2023188 | BB848823.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2023188 | BC049543.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2023188 | AF204153.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2023188 | AF204154.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2023188 | AF204155.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156876 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08767066 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161747 | - | - |
| Whole Mouse Genome G4122A | A_52_P434039 | BB848823.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P434039 | BC014288.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P434039 | BC049543.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P434039 | AF204153.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P434039 | AF204154.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P434038 | BB848823.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P434038 | BC014288.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P434038 | BC049543.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P434038 | AF204153.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P434038 | AF204157.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102757 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161747 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13315508 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13464214 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13151864 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13315515 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13105968 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13315507 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13315510 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156877 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156878 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156873 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156876 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156874 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156880 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13301532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156898 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13102116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13301542 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13151864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156878 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13301539 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13244396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156863 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156889 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10444596 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444602 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444600 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444604 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444601 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444603 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444599 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444597 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444598 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451884_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10444595 | - | - |
| Mouse Genome 430 2.0 Array | 1440450_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440450_at | - | - |
| Mouse Genome 430 2.0 Array | 1451884_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1451884_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27756_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27756_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700006572.mm.1 | NM_001110101.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006572.mm.1 | NM_001110101.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006572.mm.1 | NM_001204273.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006572.mm.1 | NM_001204273.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006572.mm.1 | NM_030597.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006572.mm.1 | NM_030597.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006572.mm.1 | TC1700000657.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451884_a_at | - | - |
| Mouse Expression Set 430 | 1451884_PM_a_at | - | - |
| Mouse Expression Set 430 | 1440450_at | - | - |
| Mouse Expression Set 430 | 1440450_PM_at | - | - |
| Murine Genome U74v2 Set | 101083_s_at | - | - |
| Murine Genome U74v2 Set | 101083_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451884_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451884_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451884_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451884_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440450_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440450_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 510082 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000510082 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2741726 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2741726 | - | - |
| mouse-6 v1.1 (Illumina) | scl50799.5.1_8-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 510082 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50799.5.1_8-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 510082 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2741726 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2800151 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2800151 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2741726 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl50799.5.1_8-S | - | - |

Lsm2
Genomic Map