Assay Details
| Gene Symbol: Abhd16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178592.3 | NP_848707.1 | 11 - 12 | 991 | 66 | |
| GenBank mRNA | BC029114.1 | - | 11 - 12 | 991 | 2648618 | 66 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Abhd16a
Genomic Map