Assay Details
| Gene Symbol: Eif4e1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033269.3 | NP_001028441.1 | 5 - 6 | 585 | 75 | |
| NM_001286178.1 | NP_001273107.1 | 5 - 6 | 514 | 75 | ||
| NM_001286179.1 | NP_001273108.1 | 5 - 6 | 517 | 75 | ||
| NR_104407.1 | 4 - 5 | 423 | 75 | |||
| GenBank mRNA | AK139577.1 | - | 4 - 5 | 823 | 75 | |
| BC139143.1 | - | 5 - 6 | 552 | 8862165 | 75 | |
| BC139144.1 | - | 5 - 6 | 549 | 8862166 | 75 | |
| DQ279473.1 | - | 5 - 6 | 580 | 75 | ||
| DQ279479.1 | - | 4 - 5 | 418 | 75 | ||
| DV653822.1 | - | 5 - 6 | 511 | 75 | ||
| DV659735.1 | - | 5 - 6 | 514 | 75 | ||
| DV662251.1 | - | 5 - 6 | 514 | 75 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eif4e1b
Genomic Map