Assay Details
| Gene Symbol: Eif4e1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033269.3 | NP_001028441.1 | 6 - 7 | 730 | 53 | |
| NM_001039683.2 | NP_001034772.1 | 6 - 7 | 692 | 53 | ||
| NM_001286178.1 | NP_001273107.1 | 6 - 7 | 659 | 53 | ||
| NM_001286179.1 | NP_001273108.1 | 6 - 7 | 662 | 53 | ||
| NM_001286180.1 | NP_001273109.1 | 6 - 7 | 621 | 53 | ||
| NR_104406.1 | 6 - 7 | 635 | 53 | |||
| NR_104407.1 | 5 - 6 | 568 | 53 | |||
| GenBank mRNA | AK139577.1 | - | 5 - 6 | 968 | 53 | |
| BC139143.1 | - | 6 - 7 | 697 | 8862165 | 53 | |
| BC139144.1 | - | 6 - 7 | 694 | 8862166 | 53 | |
| DQ279473.1 | - | 6 - 7 | 725 | 53 | ||
| DQ279474.1 | - | 6 - 7 | 670 | 53 | ||
| DQ279478.1 | - | 6 - 7 | 630 | 53 | ||
| DQ279479.1 | - | 5 - 6 | 563 | 53 | ||
| DV653822.1 | - | 6 - 7 | 656 | 53 | ||
| DV659735.1 | - | 6 - 7 | 659 | 53 | ||
| DV662251.1 | - | 6 - 7 | 659 | 53 | ||
| DV662389.1 | - | 6 - 7 | 618 | 53 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075031 | DQ279474.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075031 | AK139577.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075031 | BC139143.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075031 | BC139144.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075031 | NM_001039683.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075036 | DQ279473.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075036 | NM_001033269.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 415943 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 154679 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075031 | DQ279474.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075031 | AK139577.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075031 | BC139143.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075031 | BC139144.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075031 | NM_001039683.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075036 | DQ279473.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075036 | NM_001033269.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778818 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778816 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778826 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778808 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778827 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778797 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20778822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29051375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29051367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29051369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29051388 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29051385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29051390 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29051380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29051378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29051381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29051377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29051376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29051379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29051373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29051382 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07881134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29051465 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10989620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051342 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10687869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051332 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051351 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051347 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10985192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10771023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051383 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051343 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051377 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051357 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10985199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051345 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051370 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051348 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051344 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051367 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051381 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11001199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051375 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10985197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10723663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051385 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051376 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051369 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051354 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29051352 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11001196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10442417 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10692506 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11036454 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10442419 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10405335 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10359519 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405340 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405337 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405339 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405342 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405336 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405341 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405338 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10405334 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10359518 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300004125.mm.1 | NM_001033269.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300004125.mm.1 | NM_001039683.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300004125.mm.1 | NM_001286178.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300004125.mm.1 | NM_001286179.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300004125.mm.1 | NM_001286180.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300004125.mm.1 | NR_104406.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300004125.mm.1 | NR_104407.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300004125.mm.1 | TC1300000637.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300004128.mm.1 | NM_001033269.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300004128.mm.1 | NM_001039683.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300004128.mm.1 | NM_001286178.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300004128.mm.1 | NM_001286179.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300004128.mm.1 | NM_001286180.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300004128.mm.1 | NR_104406.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300004128.mm.1 | NR_104407.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300004128.mm.1 | TC1300000637.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300004131.mm.1 | NM_001033269.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300004131.mm.1 | NM_001039683.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300004131.mm.1 | NM_001286178.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300004131.mm.1 | NM_001286179.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300004131.mm.1 | NM_001286180.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300004131.mm.1 | NR_104406.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300004131.mm.1 | NR_104407.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300004131.mm.1 | TC1300000637.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 140753_PM_at | - | - |
| Murine Genome U74v2 Set | 140753_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254206 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3024781 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3140803 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3140803 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3024781 | - | - |

Eif4e1b
Genomic Map