Assay Details
| Gene Symbol: Pmel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021882.4 | NP_068682.2 | 5 - 6 | 663 | 64 | |
| GenBank mRNA | AK012808.1 | - | 5 - 6 | 664 | 64 | |
| AK131956.1 | - | 5 - 6 | 665 | 64 | ||
| BC075667.1 | - | 5 - 6 | 650 | 64 | ||
| BC082555.1 | - | 5 - 6 | 649 | 30541702 | 64 | |
| U14133.1 | - | 5 - 6 | 638 | 64 | ||
More Information
Set Membership: |
|

Pmel
Genomic Map