Assay Details
| Gene Symbol: Rab5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177411.4 | NP_803130.1 | 3 - 4 | 497 | 77 | |
| GenBank mRNA | AB232594.1 | - | 2 - 3 | 358 | 77 | |
| AK081251.1 | - | 3 - 4 | 493 | 77 | ||
| AK083034.1 | - | 3 - 4 | 486 | 77 | ||
| AK144551.1 | - | 3 - 4 | 619 | 77 | ||
| BC137574.1 | - | 2 - 3 | 409 | 8860593 | 77 | |
| BC137575.1 | - | 2 - 3 | 409 | 8860594 | 77 | |
| BY727964.1 | - | 3 - 4 | 498 | 77 | ||
| X84239.1 | - | 2 - 3 | 366 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rab5b
Genomic Map