Assay Details
| Gene Symbol: Cmah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111110.2 | NP_001104580.1 | 12 - 13 | 1727 | 92 | |
| NM_001284519.1 | NP_001271448.1 | 13 - 14 | 1889 | 92 | ||
| NM_001284520.1 | NP_001271449.1 | 10 - 11 | 1244 | 92 | ||
| NM_007717.5 | NP_031743.3 | 11 - 12 | 1382 | 92 | ||
| GenBank mRNA | AB061276.1 | - | 12 - 13 | 1711 | 92 | |
| AB061277.1 | - | 13 - 14 | 1873 | 92 | ||
| AK083394.1 | - | 10 - 11 | 1260 | 92 | ||
| AK145110.1 | - | 10 - 11 | 1531 | 92 | ||
| AK166486.1 | - | 11 - 12 | 1383 | 92 | ||
| BC028474.1 | - | 12 - 13 | 1737 | 92 | ||
| BC055079.1 | - | 9 - 10 | 1241 | 30077348 | 92 | |
| D21826.1 | - | 10 - 11 | 1354 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cmah
Genomic Map