Assay Details
| Gene Symbol: Cmah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111110.2 | NP_001104580.1 | 14 - 15 | 2020 | 61 | |
| NM_001284519.1 | NP_001271448.1 | 15 - 16 | 2182 | 61 | ||
| NM_001284520.1 | NP_001271449.1 | 12 - 13 | 1537 | 61 | ||
| NM_007717.5 | NP_031743.3 | 13 - 14 | 1675 | 61 | ||
| GenBank mRNA | AB061276.1 | - | 14 - 15 | 2004 | 61 | |
| AB061277.1 | - | 15 - 16 | 2166 | 61 | ||
| AK083394.1 | - | 12 - 13 | 1553 | 61 | ||
| AK145110.1 | - | 12 - 13 | 1824 | 61 | ||
| AK166486.1 | - | 13 - 14 | 1676 | 61 | ||
| BC028474.1 | - | 14 - 15 | 2030 | 61 | ||
| D21826.1 | - | 12 - 13 | 1647 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cmah
Genomic Map