Assay Details
| Gene Symbol: Zfp706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026521.4 | NP_080797.1 | 1 - 2 | 272 | 111 | |
| GenBank mRNA | AB041652.1 | - | 1 - 2 | 215 | 111 | |
| AK004076.1 | - | 1 - 2 | 71 | 111 | ||
| AK032231.1 | - | 1 - 2 | 158 | 111 | ||
| AK082392.1 | - | 1 - 2 | 176 | 111 | ||
| AK132053.1 | - | 1 - 2 | 189 | 111 | ||
| AK145319.1 | - | 1 - 2 | 150 | 111 | ||
| AK166530.1 | - | 1 - 2 | 150 | 111 | ||
| AK167791.1 | - | 1 - 2 | 260 | 111 | ||
| AK167897.1 | - | 1 - 2 | 189 | 111 | ||
| AK168303.1 | - | 1 - 2 | 189 | 111 | ||
| AK168958.1 | - | 1 - 2 | 151 | 111 | ||
| BC037600.1 | - | 1 - 2 | 197 | 111 | ||
| BC042704.1 | - | 1 - 2 | 163 | 111 | ||
| BC052459.1 | - | 1 - 2 | 272 | 111 | ||
| BC054356.1 | - | 1 - 2 | 195 | 110 | ||
| BC085477.1 | - | 1 - 2 | 156 | 111 | ||
| BC147306.1 | - | 1 - 2 | 131 | 9056379 | 111 | |
| BC147307.1 | - | 1 - 2 | 131 | 9056367 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK004076.1,AK032231.1,AK082392.1,AK132053.1,AK145319.1,AK166530.1,AK167791.1,AK167897.1,AK168303.1,AK168958.1,BC037600.1,BC042704.1,BC052459.1,BC054356.1,BC085477.1,BC147306.1,BC147307.1,NM_026521.4 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp706
Genomic Map