Assay Details
| Gene Symbol: Tcaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029930.2 | NP_084206.1 | 5 - 6 | 2062 | 89 | |
| GenBank mRNA | AK014379.1 | - | 5 - 6 | 1591 | 89 | |
| AK053509.1 | - | 5 - 6 | 2063 | 89 | ||
| AK076242.1 | - | 5 - 6 | 2008 | 89 | ||
| AK083917.1 | - | 5 - 6 | 2010 | 89 | ||
| AK132148.1 | - | 5 - 6 | 2030 | 89 | ||
| AK148593.1 | - | 5 - 6 | 2041 | 89 | ||
| AK173013.1 | - | 5 - 6 | 2044 | 89 | ||
| BC031718.1 | - | 5 - 6 | 2090 | 3156550 | 89 | |
| BC043304.1 | - | 3 - 4 | 322 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tcaf1
Genomic Map