Assay Details
| Gene Symbol: Clcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013491.2 | NP_038519.1 | 13 - 14 | 1554 | 77 | |
| GenBank mRNA | AK036735.1 | - | 13 - 14 | 1556 | 77 | |
| AK161489.1 | - | 6 - 7 | 519 | 77 | ||
| AY046403.1 | - | 13 - 14 | 1379 | 77 | ||
| AY046404.1 | - | 14 - 15 | 1458 | 77 | ||
| BC114336.1 | - | 13 - 14 | 1519 | 40048966 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clcn1
Genomic Map