Assay Details
| Gene Symbol: Clcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013491.2 | NP_038519.1 | 14 - 15 | 1668 | 117 | |
| GenBank mRNA | AK036735.1 | - | 14 - 15 | 1670 | 117 | |
| AK161489.1 | - | 7 - 8 | 633 | 117 | ||
| AY046403.1 | - | 14 - 15 | 1493 | 117 | ||
| AY046404.1 | - | 15 - 16 | 1572 | 117 | ||
| BC114336.1 | - | 14 - 15 | 1633 | 40048966 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clcn1
Genomic Map