Assay Details
| Gene Symbol: Clcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013491.2 | NP_038519.1 | 18 - 19 | 2378 | 63 | |
| GenBank mRNA | AK036735.1 | - | 18 - 19 | 2380 | 63 | |
| AK161489.1 | - | 11 - 12 | 1343 | 63 | ||
| AY046403.1 | - | 18 - 19 | 2203 | 63 | ||
| AY046404.1 | - | 19 - 20 | 2282 | 63 | ||
| BC114336.1 | - | 18 - 19 | 2343 | 40048966 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clcn1
Genomic Map