Assay Details
| Gene Symbol: B3galnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178640.2 | NP_848755.1 | 10 - 11 | 1451 | 89 | |
| GenBank mRNA | AB116655.1 | - | 10 - 11 | 1320 | 89 | |
| AK035259.1 | - | 10 - 11 | 1452 | 89 | ||
| AK041022.1 | - | 10 - 11 | 1447 | 89 | ||
| AK084275.1 | - | 10 - 11 | 1451 | 89 | ||
| AK134868.1 | - | 9 - 10 | 1214 | 89 | ||
| AK144880.1 | - | 10 - 11 | 1283 | 89 | ||
| AK151677.1 | - | 10 - 11 | 1501 | 89 | ||
| AK153362.1 | - | 10 - 11 | 1501 | 89 | ||
| AK167635.1 | - | 10 - 11 | 1452 | 89 | ||
| BC085110.1 | - | 10 - 11 | 1460 | 30735904 | 89 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

B3galnt2
Genomic Map