Assay Details
| Gene Symbol: B3galnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178640.2 | NP_848755.1 | 9 - 10 | 1294 | 135 | |
| GenBank mRNA | AB116655.1 | - | 9 - 10 | 1163 | 135 | |
| AK035259.1 | - | 9 - 10 | 1295 | 135 | ||
| AK041022.1 | - | 9 - 10 | 1290 | 135 | ||
| AK084275.1 | - | 9 - 10 | 1294 | 135 | ||
| AK085691.1 | - | 9 - 10 | 1296 | 135 | ||
| AK134868.1 | - | 8 - 9 | 1057 | 135 | ||
| AK144880.1 | - | 9 - 10 | 1125 | 135 | ||
| AK151677.1 | - | 9 - 10 | 1344 | 135 | ||
| AK153362.1 | - | 9 - 10 | 1344 | 135 | ||
| AK167635.1 | - | 9 - 10 | 1295 | 135 | ||
| BC085110.1 | - | 9 - 10 | 1303 | 30735904 | 135 | |
More Information
Updated Mapping Information
Set Membership: |
|

B3galnt2
Genomic Map