Assay Details
| Gene Symbol: Tbce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK082940.1 | - | 6 - 7 | 688 | 80 | |
| AK167383.1 | - | 6 - 7 | 676 | 80 | ||
| AK167848.1 | - | 6 - 7 | 672 | 80 | ||
| AY082332.1 | - | 6 - 7 | 642 | 80 | ||
| BC050206.1 | - | 6 - 7 | 650 | 30009463 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbce
Genomic Map