Assay Details
| Gene Symbol: Adck2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178873.3 | NP_849204.1 | 3 - 4 | 1606 | 108 | |
| GenBank mRNA | AK028557.1 | - | 3 - 4 | 1610 | 108 | |
| AK028568.1 | - | 3 - 4 | 1607 | 108 | ||
| AK161225.1 | - | 3 - 4 | 1609 | 108 | ||
| BC069944.1 | - | 3 - 4 | 1354 | 30446117 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adck2
Genomic Map