Assay Details
Gene Symbol: 1700018A04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_029439.1 | 1 - 2 | 45 | 122 | ||
GenBank mRNA | AK006076.1 | - | 1 - 2 | 38 | 122 | |
AK017122.1 | - | 1 - 2 | 46 | 122 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 11377 | Gm11377 | OTTMUSG00000000685 | -12.6kb | Anti-sense | Chr. 13 - 31564117 - 31567391 on NCBI Build GRCm38 |
Downstream | forkhead box F2 | Foxf2 |
FREAC2, Fkh20, LUN |
43.3kb | Anti-sense | Chr. 13 - 31625816 - 31631406 on NCBI Build GRCm38 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK017122.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
