Assay Details
| Gene Symbol: Ddah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190449.1 | NP_001177378.1 | 6 - 7 | 1044 | 133 | |
| GenBank mRNA | BC003328.1 | - | 6 - 7 | 994 | 3158175 | 133 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157318 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157316 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157312 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157315 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157317 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157311 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157313 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157314 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P341177 | AF004106.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P341177 | BC003328.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P341177 | AF004106.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P341177 | BC003328.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 49999 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 878110 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110181 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P341177 | AF004106.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P341177 | BC003328.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P341177 | AF004106.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P341177 | BC003328.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157326 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157323 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110181 | - | - |
| Whole Mouse Genome G4122A | A_51_P341177 | AF004106.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P341177 | BC003328.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P341177 | AF004106.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P341177 | BC003328.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131978 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110181 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13166507 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13166520 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157317 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157314 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157318 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157315 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157313 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157312 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157311 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157327 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157314 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157338 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157310 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157331 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13166533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157337 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13166509 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157321 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157316 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157332 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157334 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10444667 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444669 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444666 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444673 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444671 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444668 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444670 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444672 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416457_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10444665 | - | - |
| Mouse Genome 430 2.0 Array | 1416457_at | - | - |
| Mouse Genome 430 2.0 Array | 1416457_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 51793_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 51793_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700006689.mm.1 | NM_001190449.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006689.mm.1 | NM_001190449.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006689.mm.1 | TC1700000663.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416457_PM_at | - | - |
| Mouse Expression Set 430 | 1416457_at | - | - |
| Murine Genome U74v2 Set | 92887_PM_at | - | - |
| Murine Genome U74v2 Set | 92887_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416457_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416457_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416457_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416457_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4480551 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4480551 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004480551 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2770183 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2770183 | - | - |
| mouse-6 v1.1 (Illumina) | scl50795.8.1_75-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4480551 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50795.8.1_75-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480551 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2770183 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2770183 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl50795.8.1_75-S | - | - |

Ddah2
Genomic Map