Assay Details
| Gene Symbol: Lsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204273.1 | NP_001191202.1 | 2 - 3 | 307 | 59 | |
| GenBank mRNA | AF204154.1 | - | 2 - 3 | 278 | 59 | |
| AF204155.1 | - | 2 - 3 | 278 | 59 | ||
| AF204156.1 | - | 2 - 3 | 228 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF204154.1, AF204155.1 transcripts.
Set Membership: |
|

Lsm2
Genomic Map