Assay Details
| Gene Symbol: Ppt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002301.1 | - | 3 - 4 | 568 | 96 | |
| BC013462.1 | - | 3 - 4 | 567 | 4238225 | 96 | |
| BC052330.1 | - | 4 - 5 | 541 | 96 | ||
| CX217389.1 | - | 4 - 5 | 504 | 96 | ||
| CX565661.1 | - | 4 - 5 | 499 | 96 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ppt2
Genomic Map