Assay Details
| Gene Symbol: Galnt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144908.3 | NP_659157.1 | 4 - 5 | 757 | 93 | |
| GenBank mRNA | AF495528.1 | - | 3 - 4 | 629 | 93 | |
| AK088014.1 | - | 4 - 5 | 746 | 93 | ||
| AK161524.1 | - | 4 - 5 | 667 | 93 | ||
| BC011428.1 | - | 4 - 5 | 733 | 2631696 | 93 | |
| BC021504.1 | - | 4 - 5 | 758 | 5346678 | 93 | |
| BC031150.1 | - | 3 - 4 | 655 | 93 | ||
| BC034184.1 | - | 3 - 4 | 657 | 4974216 | 93 | |
| BC034185.1 | - | 3 - 4 | 709 | 4974695 | 93 | |
| BC036143.1 | - | 3 - 4 | 658 | 4974645 | 93 | |
| BC036145.1 | - | 3 - 4 | 651 | 4975474 | 93 | |
| Y12435.1 | - | 3 - 4 | 580 | 93 | ||
More Information
Set Membership: |
|

Galnt11
Genomic Map