Assay Details
| Gene Symbol: Cdk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007668.3 | NP_031694.1 | 4 - 5 | 293 | 72 | |
| GenBank mRNA | AK051790.1 | - | 4 - 5 | 293 | 72 | |
| BC052007.1 | - | 4 - 5 | 267 | 5706932 | 72 | |
| D29678.1 | - | 4 - 5 | 302 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk5
Genomic Map