Assay Details
| Gene Symbol: Smarcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025891.3 | NP_080167.3 | 8 - 9 | 1083 | 130 | |
| GenBank mRNA | AK005094.1 | - | 8 - 9 | 1030 | 130 | |
| AK019374.1 | - | 1 - 2 | 164 | 130 | ||
| AK158019.1 | - | 1053 | 130 | |||
| BC013122.1 | - | 8 - 9 | 1120 | 3586276 | 130 | |
| BC060525.1 | - | 8 - 9 | 982 | 6817219 | 130 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smarcd3
Genomic Map