Assay Details
| Gene Symbol: Nub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF534114.1 | - | 9 - 10 | 1092 | 114 | |
| AK018166.1 | - | 9 - 10 | 1901 | 114 | ||
| AK086301.1 | - | 10 - 11 | 1302 | 114 | ||
| AK134581.1 | - | 10 - 11 | 1146 | 114 | ||
| AK153400.1 | - | 10 - 11 | 1130 | 114 | ||
| BC005640.1 | - | 10 - 11 | 1131 | 114 | ||
| BC018178.1 | - | 10 - 11 | 1117 | 114 | ||
| BC037451.1 | - | 9 - 10 | 1121 | 4018648 | 114 | |
| U27462.1 | - | 10 - 11 | 1339 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nub1
Genomic Map