Assay Details
| Gene Symbol: Nub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF534114.1 | - | 11 - 12 | 1390 | 74 | |
| AK018166.1 | - | 11 - 12 | 2199 | 74 | ||
| AK086301.1 | - | 12 - 13 | 1600 | 74 | ||
| AK134581.1 | - | 12 - 13 | 1444 | 74 | ||
| BC037451.1 | - | 11 - 12 | 1419 | 4018648 | 74 | |
| U27462.1 | - | 12 - 13 | 1637 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nub1
Genomic Map