Assay Details
| Gene Symbol: Nub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF534114.1 | - | 3 - 4 | 343 | 125 | |
| AK086301.1 | - | 4 - 5 | 553 | 125 | ||
| AK134581.1 | - | 4 - 5 | 397 | 125 | ||
| AK153400.1 | - | 4 - 5 | 381 | 125 | ||
| BC005640.1 | - | 4 - 5 | 382 | 125 | ||
| BC018178.1 | - | 4 - 5 | 368 | 125 | ||
| BC037451.1 | - | 3 - 4 | 372 | 4018648 | 125 | |
| U27462.1 | - | 4 - 5 | 590 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nub1
Genomic Map