Assay Details
| Gene Symbol: Nub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF534114.1 | - | 7 - 8 | 801 | 127 | |
| AK018166.1 | - | 7 - 8 | 1610 | 127 | ||
| AK086301.1 | - | 8 - 9 | 1011 | 127 | ||
| AK134581.1 | - | 8 - 9 | 855 | 127 | ||
| AK153400.1 | - | 8 - 9 | 839 | 127 | ||
| BC005640.1 | - | 8 - 9 | 840 | 127 | ||
| BC018178.1 | - | 8 - 9 | 826 | 127 | ||
| BC037451.1 | - | 7 - 8 | 830 | 4018648 | 127 | |
| U27462.1 | - | 8 - 9 | 1048 | 127 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nub1
Genomic Map