Assay Details
| Gene Symbol: Pbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017463.2 | NP_059491.1 | 5 - 6 | 950 | 65 | |
| GenBank mRNA | AF020198.1 | - | 5 - 6 | 952 | 65 | |
| AK134357.1 | - | 5 - 6 | 871 | 65 | ||
| AK141763.1 | - | 874 | 65 | |||
| AK159623.1 | - | 5 - 6 | 1159 | 65 | ||
| BC010287.1 | - | 6 - 7 | 1062 | 2599030 | 65 | |
| BC062147.1 | - | 5 - 6 | 1126 | 6505014 | 65 | |
| BC087615.1 | - | 5 - 6 | 930 | 4978305 | 65 | |
| CT010391.1 | - | 5 - 6 | 867 | 65 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pbx2
Genomic Map