Assay Details
| Gene Symbol: Nupl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153092.4 | NP_694732.3 | 2 - 3 | 464 | 84 | |
| GenBank mRNA | AB067574.1 | - | 2 - 3 | 389 | 84 | |
| AK077066.1 | - | 2 - 3 | 431 | 84 | ||
| AK078478.1 | - | 2 - 3 | 465 | 84 | ||
| BC033270.1 | - | 2 - 3 | 406 | 3981211 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nupl2
Genomic Map