Assay Details
| Gene Symbol: Nupl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153092.4 | NP_694732.3 | 3 - 4 | 556 | 65 | |
| GenBank mRNA | AK078478.1 | - | 3 - 4 | 557 | 65 | |
| BC033270.1 | - | 3 - 4 | 498 | 3981211 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nupl2
Genomic Map