Assay Details
| Gene Symbol: Psmc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011188.3 | NP_035318.1 | 9 - 10 | 1159 | 128 | |
| GenBank mRNA | AK005083.1 | - | 8 - 9 | 782 | 128 | |
| AK076874.1 | - | 9 - 10 | 1161 | 128 | ||
| AK146919.1 | - | 8 - 9 | 800 | 128 | ||
| AK153413.1 | - | 8 - 9 | 803 | 128 | ||
| AK168548.1 | - | 8 - 9 | 800 | 128 | ||
| BC005462.1 | - | 8 - 9 | 809 | 3590739 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psmc2
Genomic Map