Assay Details
| Gene Symbol: Hgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289458.1 | NP_001276387.1 | 14 - 15 | 1740 | 93 | |
| NM_001289459.1 | NP_001276388.1 | 14 - 15 | 1729 | 93 | ||
| NM_001289461.1 | NP_001276390.1 | 13 - 14 | 1547 | 93 | ||
| NM_010427.5 | NP_034557.3 | 14 - 15 | 1712 | 93 | ||
| GenBank mRNA | BC119228.1 | - | 14 - 15 | 1599 | 8733977 | 93 |
| BC144764.1 | - | 14 - 15 | 1611 | 93 | ||
| D10212.1 | - | 13 - 14 | 1535 | 93 | ||
| D10213.1 | - | 13 - 14 | 1520 | 93 | ||
| S71816.1 | - | 13 - 14 | 1535 | 93 | ||
| X72307.1 | - | 13 - 14 | 1596 | 93 | ||
| X84046.1 | - | 13 - 14 | 1575 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hgf
Genomic Map