Assay Details
| Gene Symbol: Hgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289458.1 | NP_001276387.1 | 2 - 3 | 297 | 123 | |
| NM_001289459.1 | NP_001276388.1 | 2 - 3 | 286 | 123 | ||
| NM_001289460.1 | NP_001276389.1 | 2 - 3 | 297 | 123 | ||
| NM_001289461.1 | NP_001276390.1 | 1 - 2 | 119 | 123 | ||
| NM_010427.5 | NP_034557.3 | 2 - 3 | 269 | 123 | ||
| GenBank mRNA | AF042856.1 | - | 1 - 2 | 98 | 123 | |
| AK042121.1 | - | 2 - 3 | 215 | 123 | ||
| AK052289.1 | - | 2 - 3 | 298 | 123 | ||
| AK082461.1 | - | 1 - 2 | 269 | 123 | ||
| AK142159.1 | - | 2 - 3 | 215 | 123 | ||
| BC119228.1 | - | 2 - 3 | 156 | 8733977 | 123 | |
| BC144764.1 | - | 2 - 3 | 167 | 123 | ||
| D10212.1 | - | 1 - 2 | 92 | 123 | ||
| D10213.1 | - | 1 - 2 | 92 | 123 | ||
| S71816.1 | - | 1 - 2 | 92 | 123 | ||
| X72307.1 | - | 1 - 2 | 153 | 123 | ||
| X84046.1 | - | 1 - 2 | 132 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hgf
Genomic Map