Assay Details
| Gene Symbol: Gpm6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177955.1 | NP_001171426.1 | 2 - 3 | 648 | 55 | |
| NM_001177956.1 | NP_001171427.1 | 2 - 3 | 648 | 55 | ||
| NM_001177957.1 | NP_001171428.1 | 2 - 3 | 648 | 55 | ||
| GenBank mRNA | AF254870.1 | - | 2 - 3 | 509 | 55 | |
| AF254874.1 | - | 2 - 3 | 509 | 55 | ||
| AF254875.1 | - | 2 - 3 | 509 | 55 | ||
| BQ745381.1 | - | 2 - 3 | 483 | 55 | ||
More Information
Set Membership: |
|

Gpm6b
Genomic Map