Assay Details
| Gene Symbol: Ctps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168568.1 | NP_001162040.1 | 15 - 16 | 1673 | 111 | |
| NM_001168569.1 | NP_001162041.1 | 15 - 16 | 1730 | 111 | ||
| NM_018737.5 | NP_061207.1 | 15 - 16 | 1805 | 111 | ||
| NR_033143.1 | 15 - 16 | 1695 | 111 | |||
| GenBank mRNA | AK032018.1 | - | 15 - 16 | 1532 | 111 | |
| AK052854.1 | - | 15 - 16 | 1721 | 111 | ||
| AK084595.1 | - | 15 - 16 | 1602 | 111 | ||
| AK085381.1 | - | 15 - 16 | 1581 | 111 | ||
| AK150966.1 | - | 15 - 16 | 1672 | 111 | ||
| AK151597.1 | - | 15 - 16 | 1672 | 111 | ||
| BC003257.1 | - | 15 - 16 | 1513 | 3500778 | 111 | |
| EF410142.1 | - | 404 | 111 | |||
| U49385.1 | - | 15 - 16 | 1695 | 111 | ||
More Information
Set Membership: |
|

Ctps2
Genomic Map