Assay Details
| Gene Symbol: Ctps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168568.1 | NP_001162040.1 | 16 - 17 | 1770 | 75 | |
| NM_001168569.1 | NP_001162041.1 | 16 - 17 | 1827 | 75 | ||
| NM_018737.5 | NP_061207.1 | 16 - 17 | 1902 | 75 | ||
| NR_033143.1 | 16 - 17 | 1792 | 75 | |||
| GenBank mRNA | AK032018.1 | - | 16 - 17 | 1629 | 75 | |
| AK052854.1 | - | 16 - 17 | 1818 | 75 | ||
| AK084595.1 | - | 16 - 17 | 1699 | 75 | ||
| AK085381.1 | - | 16 - 17 | 1678 | 75 | ||
| AK150966.1 | - | 16 - 17 | 1769 | 75 | ||
| AK151597.1 | - | 16 - 17 | 1769 | 75 | ||
| BC003257.1 | - | 16 - 17 | 1610 | 3500778 | 75 | |
| EF410142.1 | - | 501 | 75 | |||
| U49385.1 | - | 16 - 17 | 1792 | 75 | ||
More Information
Set Membership: |
|

Ctps2
Genomic Map