Assay Details
Gene Symbol: Prl7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164058.1 | NP_001157530.1 | 3 - 4 | 295 | 112 | |
NM_008930.2 | NP_032956.1 | 3 - 4 | 292 | 112 | ||
GenBank mRNA | AF011381.1 | - | 3 - 4 | 284 | 112 | |
AF020525.1 | - | 3 - 4 | 266 | 112 | ||
AK028371.1 | - | 3 - 4 | 293 | 112 | ||
AK141309.1 | - | 3 - 4 | 296 | 112 | ||
AK146050.1 | - | 3 - 4 | 296 | 112 | ||
BB611867.1 | - | 3 - 4 | 293 | 112 | ||
BC051671.1 | - | 3 - 4 | 277 | 30018275 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
