Assay Details
| Gene Symbol: Prl8a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271378.1 | NP_001258307.1 | 4 - 5 | 450 | 103 | |
| NM_011167.3 | NP_035297.1 | 4 - 5 | 447 | 103 | ||
| NR_073166.1 | 4 - 5 | 372 | 103 | |||
| GenBank mRNA | AF090140.1 | - | 4 - 5 | 356 | 103 | |
| AK005435.1 | - | 4 - 5 | 411 | 103 | ||
| BC099469.1 | - | 4 - 5 | 393 | 30787452 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2269459 | BC099469.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2269459 | AK005478.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2269459 | NM_011167.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2269459 | AF090140.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2269459 | AK005435.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303122 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 687999 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 660987 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 127128 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115503 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2269459 | BC099469.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2269459 | AK005478.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2269459 | NM_011167.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2269459 | AF090140.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2269459 | AK005435.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902259 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115503 | - | - |
| Whole Mouse Genome G4122A | A_51_P430855 | BC099469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430855 | AK005478.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430855 | NM_011167.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430855 | AF090140.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430855 | AK005435.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100994 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102081 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115503 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10692451 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10692453 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10692452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902215 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10472806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902268 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902245 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10479461 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10622705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902202 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10716163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902233 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902275 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10622703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902239 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10819749 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10408384 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408382 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408385 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408383 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408381 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408380 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418418_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408379 | - | - |
| Mouse Genome 430 2.0 Array | 1418418_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1418418_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19112_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19112_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300010493.mm.1 | NM_001271378.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010493.mm.1 | NM_001271379.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010493.mm.1 | NM_011167.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010493.mm.1 | NR_073166.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010493.mm.1 | TC1300001690.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300010497.mm.1 | NM_001271378.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010497.mm.1 | NM_001271379.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010497.mm.1 | NM_011167.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010497.mm.1 | NR_073166.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010497.mm.1 | TC1300001690.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418418_PM_a_at | - | - |
| Mouse Expression Set 430 | 1418418_a_at | - | - |
| Murine Genome U74v2 Set | 99530_PM_at | - | - |
| Murine Genome U74v2 Set | 99530_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418418_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418418_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418418_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418418_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 816578 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3610114 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3610114 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003610114 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2716897 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2716897 | - | - |
| mouse-6 v1.1 (Illumina) | scl44172.6.1_34-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3610114 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl44172.6.1_34-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3610114 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2716897 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2716897 | - | - |

Prl8a6
Genomic Map