Assay Details
| Gene Symbol: Prl8a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289919.1 | NP_001276848.1 | 5 - 6 | 592 | 108 | |
| NM_010088.2 | NP_034218.1 | 5 - 6 | 595 | 108 | ||
| GenBank mRNA | AF011385.1 | - | 5 - 6 | 579 | 108 | |
| AF015729.1 | - | 5 - 6 | 589 | 108 | ||
| AK131668.1 | - | 5 - 6 | 593 | 108 | ||
| AK146049.1 | - | 5 - 6 | 596 | 108 | ||
| BC061150.1 | - | 5 - 6 | 592 | 30317747 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057112 | AF015729.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057112 | AK131668.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057112 | AK146049.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057112 | AF011385.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057112 | NM_010088.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316314 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 210741 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 211736 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 27089 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 62902 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P176532 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057112 | AF015729.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057112 | AK131668.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057112 | AK146049.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057112 | AF011385.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057112 | NM_010088.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901889 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P176532 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07818503 | - | - |
| Whole Mouse Genome G4122A | A_51_P497044 | AF015729.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497044 | AK131668.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497044 | AK146049.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497044 | AF011385.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497044 | BC061150.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121766 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127218 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P176532 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10983079 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11062032 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10983077 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10745713 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10983081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901914 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10674509 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901920 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10989941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901880 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901921 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901905 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901919 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901914 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10835901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901899 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901868 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10996961 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28901925 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10404317 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404322 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404318 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404321 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404319 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404320 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448608_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10404316 | - | - |
| Mouse Genome 430 2.0 Array | 1448608_at | - | - |
| Mouse Genome 430 2.0 Array | 1448608_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13529_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13529_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300001876.mm.1 | NM_010088.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001876.mm.1 | TC1300000303.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300001880.mm.1 | NM_010088.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001880.mm.1 | TC1300000303.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300001879.mm.1 | NM_010088.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001879.mm.1 | TC1300000303.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300001881.mm.1 | NM_010088.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001881.mm.1 | TC1300000303.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300001882.mm.1 | NM_010088.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001882.mm.1 | TC1300000303.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300001878.mm.1 | NM_010088.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001878.mm.1 | TC1300000303.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448608_at | - | - |
| Mouse Expression Set 430 | 1448608_PM_at | - | - |
| Murine Genome U74v2 Set | 98529_at | - | - |
| Murine Genome U74v2 Set | 98529_PM_at | - | - |
| Murine Genome U74v2 Set | 167123_PM_i_at | - | - |
| Murine Genome U74v2 Set | 167123_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448608_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448608_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448608_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448608_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 666249 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5900278 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5900278 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005900278 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225722 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225722 | - | - |
| mouse-6 v1.1 (Illumina) | scl44963.6.1_112-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5900278 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl44963.6.1_112-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5900278 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225722 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225722 | - | - |

Prl8a2
Genomic Map