Assay Details
| Gene Symbol: Cdkal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018346.1 | - | 1 - 2 | 76 | 121 | |
| AK041864.1 | - | 1 - 2 | 72 | 121 | ||
| AK045506.1 | - | 1 - 2 | 96 | 121 | ||
| AK082629.1 | - | 1 - 2 | 79 | 121 | ||
| AK158546.1 | - | 1 - 2 | 69 | 121 | ||
| AK169761.1 | - | 1 - 2 | 76 | 121 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK041864.1, AK045506.1, AK082629.1, AK169761.1 transcripts.
Set Membership: |
|

Cdkal1
Genomic Map