Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600000315.mm.1 |
TC0600000027.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600000315.mm.1 |
NM_001191023.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000315.mm.1 |
NM_001191023.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000315.mm.1 |
NM_001191025.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000315.mm.1 |
NM_001191025.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000315.mm.1 |
NM_001191026.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000315.mm.1 |
NM_001191026.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000315.mm.1 |
NM_001191027.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000315.mm.1 |
NM_001191027.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000315.mm.1 |
NM_010063.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000315.mm.1 |
NM_010063.4 |
Refseq |