Assay Details
| Gene Symbol: Pon2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183308.2 | NP_899131.1 | 3 - 4 | 291 | 55 | |
| GenBank mRNA | AK007350.1 | - | 3 - 4 | 259 | 55 | |
| AK146311.1 | - | 3 - 4 | 259 | 55 | ||
| AK152351.1 | - | 3 - 4 | 242 | 55 | ||
| AK171926.1 | - | 3 - 4 | 249 | 55 | ||
| AK210313.1 | - | 2 - 3 | 114 | 55 | ||
| AK213462.1 | - | 2 - 3 | 114 | 55 | ||
| BC021887.1 | - | 3 - 4 | 248 | 3154583 | 55 | |
| BC037140.1 | - | 3 - 4 | 233 | 5027719 | 55 | |
| BC055896.1 | - | 3 - 4 | 291 | 3482584 | 55 | |
| BC062200.1 | - | 3 - 4 | 282 | 6399484 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pon2
Genomic Map